Thiophenol 99%, AcroSeal™
|
Danger
|
AC43072-1000
AC43072-8000
103528-792EA
172
USD
103528-792
103528-790
Thiophenol 99%, AcroSeal™
Thiophenol
Formula:
C₆H₆S MW: 110.18 g/mol Boiling Pt: 167…169 °C (760 mmHg) Melting Pt: –15 °C Density: 1.078 g/cm³ Flash Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004826 CAS Number: 108-98-5 UN: 2337 ADR: 6.1,I |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5880 to 1.5900 (20 °C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...