Thiophenol 99%
|
Danger
|
AC13887-0010
AC13887-0050
103525-530EA
642
USD
103525-530
103525-528
Thiophenol 99%
Thiophenol
Formula:
C₆H₆S MW: 110.18 g/mol Boiling Pt: 167…169 °C (760 mmHg) Melting Pt: –15 °C Density: 1.078 g/cm³ Flash Pt: 50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004826 CAS Number: 108-98-5 EINECS: 203-635-3 UN: 2337 ADR: 6.1, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...