5-Thiazolemethanol 95%
|
Danger
|
Synonyms:
Thiazol-5-ylmethanol
, Thiazole-5-methanol
, 5-(Hydroxymethyl)thiazole
AC45555-0050
AC45555-0250
103517-628EA
40.8
USD
103517-628
103517-630
5-Thiazolemethanol 95%
5-Thiazolemethanol
Formula:
C₄H₅NOS MW: 115.15 g/mol Density: 1.32 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD04115732 CAS Number: 38585-74-9 |
Specification Test Results
Appearance | Clear light yellow to orange Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5670 to 1.5710 (20 °C, 589 nm) |
GC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...