Tetrakis(triphenylphosphine)palladium(0) ≥99.9% (trace metals basis)

Supplier: ACROS ORGANICS MS

Warning

Synonyms: Pd(PPh3)4

AC44140-0010 AC44140-0050
103518-930EA 67.02 USD
103518-930 103518-932
Tetrakis(triphenylphosphine)palladium(0) ≥99.9% (trace metals basis)
Tetrakis(triphenylphosphine)palladium (0)
Formula: C₇₂H₆₀P₄Pd
MW: 1155,58 g/mol
Melting Pt: 103 …107 °C (dec.)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9

Order Now

Specification Test Results

Appearance Yellow to golden Crystalline powder or crystals
Infrared spectrum Conforms
Total trace metal impurities ≤0.1 %
Palladium (Pd) 8.8 to 9.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR