Tetrahydropyran-4-sulfonyl chloride 97%

Danger

Synonyms: Tetrahydro-2H-pyran-4-sulphonyl chloride2H-Pyran-4-sulphonylchloride,tetrahydro-oxane-4-sulfonyl chlorideTetrahydro-2H-pyran-4-sulfonyl chloride2H-Pyran-4-sulfonylchloride,tetrahydro-oxane-4-sulphonyl chloride

AC42944-0010 AC42944-0050
103528-776EA 673 USD
103528-776 103524-362
Tetrahydropyran-4-sulfonyl chloride 97%
Tetrahydropyran-4-sulfonyl chloride
Formula: C₅H₉ClO₃S
MW: 184.634 g/mol
Melting Pt: 36…40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07787616
CAS Number: 338453-21-7
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to light brown Low melting solid
Infrared spectrum Conforms
Melting point 35 to 41 °C
GC ≥96.0 %
Residue after evaporation ≤0.0025 %
Titrable acid ≤0.0005 meq/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR