Tetrahydropyran-4-carboxylic acid 97%
|
Warning
|
Synonyms:
Tetrahydropyran-4-yl-carboxylic acid
AC45672-0250
AC45672-0050
103517-514EA
60.6
USD
103517-514
103517-516
Tetrahydropyran-4-carboxylic acid 97%
Tetrahydropyran-4-carboxylic acid
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 115 °C (20 mmHg) Melting Pt: 87…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031016 CAS Number: 5337-03-1 |
Specification Test Results
Appearance | White to yellow to brown Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 87 to 91 °C |
Water | ≤1.5 % |
Titration with NaOH | 96.5 to 103.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...