Tetrabutylammonium hydroxide-30-hydrate ≥95%

Supplier: ACROS ORGANICS MS

Danger

Synonyms: N,N,N-Tributylbutan-1-aminium hydroxide--water (1/1/30)TETRABUTYLAMMONIUM TRIACONTAHYDRATE HYDROXIDEMolPort-039-140-470

AC43483-1000 AC43483-0250
103524-442EA 177.8 USD
103524-442 103524-440
Tetrabutylammonium hydroxide-30-hydrate ≥95%
Tetrabutylammonium hydroxide-30-hydrate
Formula: C₁₆H₉₇NO₃₁
MW: 799,928 g/mol
Melting Pt: 27…30 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00149573
CAS Number: 147741-30-8
UN: 3263
ADR: 8,II

Order Now

Specification Test Results

Appearance Transparent white Crystals or crystalline powder
Melting point 26°C to 30°C
Titration with HCl ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR