Tetrabutylammonium chloride xhydrate 96%

Warning

Synonyms: Tetrabutyl ammonium chloride xhydrateTetrabutylammonium chloride hydrateTBACTetra-n-butylammonium chloride hydrate

AC43206-0050
103515-400EA 26.5 USD
103515-400
Tetrabutylammonium chloride xhydrate 96%
Tetrabutylammonium chloride hydrate
Formula: C₁₆H₃₆ClN·xH₂O
MW: 277.92 g/mol (anhy)
Melting Pt: 37…39 °C
Storage Temperature: Ambient
MDL Number: MFCD00011635
CAS Number: 37451-68-6

Order Now

Specification Test Results

Appearance Off-white Low melting solid
Infrared spectrum Conforms
Titration Argentometric ≥95.0 % (On dried substance)
Water ≤10.0 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR