tert-Butyl methyl ether for analysis

Danger

Synonyms: Methyl tert-butyl etherMethyl 1,1-dimethylethyl etherMethyl t-butyl etherEther, tert-butyl methyl2-Methyl-2-methoxypropane2-Methoxy-2-methylpropaneMTBE

AC39048-0010 AC39048-0025
103528-398EA 138.46 USD
103528-398 103528-396
tert-Butyl methyl ether for analysis
tert-Butyl methyl ether
Formula: (CH₃)₃COCH₃
MW: 88.15 g/mol
Boiling Pt: 54…56 °C (760 mmHg)
Melting Pt: –110 °C
Density: 0.7404 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
UN: 2398
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/77/75/2297775.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.05 %
GC ≥99.5 %
Residue after evaporation ≤0.005 %
Acidity ≤0.0005 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Boron (B) ≤0.02 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Peroxides ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR