tert-Butanol 99.5% for analysis

Danger

Synonyms: Trimethylcarbinoltert-Butyl alcohol2-Methyl-2-propanol

AC44701-0025 AC44701-0010
103528-926EA 113.31 USD
103528-926 103528-928
tert-Butanol 99.5% for analysis
tert-Butanol
Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 83 °C (760 mmHg)
Melting Pt: 25…25.5 °C
Density: 0.78 g/cm³
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
UN: 1120
ADR: 3,II
https://us.vwr.com/stibo/web/std.lang.all/12/89/2301289.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3860 to 1.3880 (20 °C, 589 nm)
Water ≤0.1 %
GC ≥99.4 %
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Acidity ≤0.0005 meq/g
Alkalinity ≤0.0002 meq/g
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Boron (B) ≤0.02 ppm
Cadmium (Cd) ≤0.05 ppm
Calcium (Ca) ≤0.5 ppm
Chromium (Cr) ≤0.02 ppm
Cobalt (Co) ≤0.02 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Manganese (Mn) ≤0.02 ppm
Nickel (Ni) ≤0.02 ppm
Tin (Sn) ≤0.1 ppm
Zinc (Zn) ≤0.1 ppm
Carbonyl compounds ≤0.01 % (as butyraldehyde)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR