TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulfonic acid) for biochemistry
|
Warning
|
Synonyms:
2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulphonic acid
, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulphonic acid
, 3-[N-Tris(hydroxymethyl)methylamino]-2-hydroxypropanesulfonic acid
, 2-Hydroxy-3-{[2-hydroxy-1,1-bis(hydroxymethyl)-ethyl]amino}-1-propanesulfonic acid
AC32781-5000
AC32781-0250
AC32781-1000
103466-538EA
250
USD
103466-538
103466-536
103466-534
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulfonic acid) for biochemistry
TAPSO (2-Hydroxy-3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid)
Formula:
C₇H₁₇NO₇S MW: 259.28 g/mol Melting Pt: 220…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038352 CAS Number: 68399-81-5 EINECS: 269-993-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...