Sulfuryl chloride 98.5%
|
Danger
|
Synonyms:
Sulphuryl dichloride
, Sulfuryl dichloride
AC16945-0010
AC16945-0025
103526-264EA
172
USD
103526-264
103526-262
Sulfuryl chloride 98.5%
Sulphuryl chloride
Formula:
Cl₂O₂S MW: 134.97 g/mol Boiling Pt: 69.1 °C (760 mmHg) Melting Pt: –54.1 °C Density: 1.667 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011451 CAS Number: 7791-25-5 EINECS: 232-245-6 UN: 1834 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...