Sulfur trioxide trimethylamine complex 95%

Danger

AC15935-1000 AC15935-5000
103526-022EA 328 USD
103526-022 103526-020
Sulfur trioxide trimethylamine complex 95%
Sulphur trioxide trimethylamine complex
Formula: C₃H₉NO₃S
Melting Pt: 232 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012421
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR