Sulfamethoxypyridazine
|
Danger
|
Synonyms:
4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulfonamide
, 4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide
AC43647-0250
AC43647-0050
103466-520EA
40.2
USD
103466-520
103466-522
Sulfamethoxypyridazine
Sulphamethoxypyridazine
Formula:
C₁₁H₁₂N₄O₃S MW: 280.3 g/mol Melting Pt: 182…183 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00057372 CAS Number: 80-35-3 |
Specification Test Results
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 180 to 184 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...