Sulfaguanidine 98%

Warning

Synonyms: 4-Amino-N-carbamimidoylbenzenesulfonamide4-Amino-N-carbamimidoylbenzenesulphonamide4-Amino-N-amidinobenzenesulphonamide4-Aminobenzenesulfonylguanidine4-Amino-N-amidinobenzenesulfonamide4-Aminobenzenesulphonylguanidine

AC43130-1000 AC43130-0250
103515-366EA 41.4 USD
103515-366 103515-364
Sulfaguanidine 98%
Sulfaguanidine
Formula: C₇H₁₀N₄O₂S
MW: 214.24 g/mol
Melting Pt: 190.5…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00038136
CAS Number: 57-67-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 189 to 193 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR