Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™

Danger

Synonyms: Sodium tert-pentoxide, 95%tert-Amyl alcohol sodium saltSodium tert-pentoxide

AC25253-1000 AC25253-8000
103527-110EA 76.05 USD
103527-110 103527-112
Sodium-tert-pentyloxide 30% in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal™
Sodium-tert-pentyloxide
Formula: C₅H₁₁NaO
MW: 110.132 g/mol
Density: 0.91 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00064245
CAS Number: 14593-46-5
UN: 2924
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR