Sodium pivalate hydrate 99%
|
Warning
|
Synonyms:
Propanoic acid, 2,2-dimethyl-, sodium salt, hydrate
, Sodium trimethylacetate hydrate
, Trimethylacetic acid sodium salt hydrate
AC36471-0100
AC36471-0500
103512-746EA
64.8
USD
103512-746
103512-748
Sodium pivalate hydrate 99%
Sodium pivalate hydrate
Formula:
C₅H₁₁NaO₃ MW: 142.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150782 CAS Number: 143174-36-1 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤7 % (K.F.) |
Titration with HClO4 | ≥98.5 % (On dry substance) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...