Trisodium phosphate dodecahydrate 98% for biochemistry

Warning

Synonyms: Sodium Phosphate tribasic 12-HydrateSodium phosphate tribasic dodecahydrateTrisodium phosphateSodium phosphate dodecahydrate

AC44796-0025 AC44796-5000
103516-976EA 509 USD
103516-976 103516-978
Trisodium phosphate dodecahydrate 98% for biochemistry
tri-Sodium phosphate dodecahydrate
Formula: Na₃PO₄·12H₂O
MW: 380.12 g/mol
Melting Pt: 75 °C
Density: 1.62 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149198
CAS Number: 10101-89-0

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder
UV (0.5 M in water)
UV at 260 nm A: ≤0.035
UV at 280 nm A: ≤0.025
Titration excess HCl 97.5 to 102.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.01 %
Cadmium (Cd) ≤0.0005 %
Calcium (Ca) ≤0.005 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Nickel (Ni) ≤0.0005 %
Nitrogen (N) ≤0.001 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤0.001 %
Free alkali (NaOH) ≤2.5 %
Insoluble matter ≤0.01 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR