Sodium nitrate, Extra Pure

Warning

AC43609-0050 AC43609-0010
103524-454EA 48.5 USD
103524-454 103524-456
Sodium nitrate, Extra Pure
Sodium nitrate
Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (760 mmHg)
Melting Pt: 306 °C
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
UN: 1498
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Colorless to white Crystalline powder or crystals
Loss on drying ≤2 % (105 °C)
pH 5.5 to 8.3 (5 % in water)
Titration after Ion exchange ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR