Sodium chloride 99.5% for biochemistry

Synonyms: SaltHaliteSea saltRock saltSaline

AC44730-2500 AC44730-0050 AC44730-0010
103516-876EA 34.31 USD
103516-876 103516-872 103516-874
Sodium chloride 99.5% for biochemistry
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C (760 mmHg)
Melting Pt: 801 °C
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance White Powder
Titration Argentometric ≥99.4 %
Solubility (0.1 M in water) Clear colorless
UV (0.1 M in water)
UV at 260 nm A: ≤0.01
UV at 280 nm A: ≤0.01
Color scale ≤15 APHA (0.1 M in water)
pH 5.0 to 9.0
Heavy metals (as Pb) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR