Sodium chloride 99.5% for HPLC

Synonyms: SaltHaliteSea saltRock saltSaline

AC44621-2500
103516-800EA 156 USD
103516-800
Sodium chloride 99.5% for HPLC
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1461 °C (760 mmHg)
Melting Pt: 801 °C
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.4 %
UV (10 % in water)
UV at 220 nm A: ≤0.05
UV at 230 nm A: ≤0.03
UV at 240 nm A: ≤0.02
UV at 260 nm A: ≤0.01
UV at 500 nm A: ≤0.01
pH 5.0 to 9.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR