Sodium carbonate decahydrate 99+% for biochemistry

Warning

AC12502-0025 AC12502-5000
103516-444EA 49.1 USD
103516-444 103516-442
Sodium carbonate decahydrate 99+% for biochemistry
Sodium carbonate decahydrate
Formula: Na₂CO₃·10H₂O
MW: 286.14 g/mol
Melting Pt: 34 °C
Density: 1.46 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149178
CAS Number: 6132-02-1

Order Now

Specification Test Results

Appearance White Crystals
Solubility Clear colorless
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.03
UV (0.5 M in water)
pH (0.5 M in water) Approx. 12.0
Titration with HCl ≥99.0 %
Chloride (Cl) ≤0.001 %
PO4 and SiO2 (as SiO2) ≤0.003 %
Aluminium (Al) ≤0.0005 %
Arsenic (As) ≤0.0002 %
Calcium (Ca) ≤0.002 %
Copper (Cu) ≤0.0005 %
Iron (Fe) ≤0.0002 %
Magnesium (Mg) ≤0.0002 %
Potassium (K) ≤0.005 %
Zinc (Zn) ≤0.0005 %
Heavy metals (as Pb) ≤5 ppm
Total sulfur (as SO4) ≤0.003 %
Total nitrogen (as N) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR