Sodium carbonate 99.999% (trace metals basis)

Warning

Synonyms: Na-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

AC43680-0250 AC43680-0050
103518-070EA 51.9 USD
103518-070 103518-072
Sodium carbonate 99.999% (trace metals basis)
Sodium carbonate
Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600 °C (760 mmHg)
Melting Pt: 851 °C
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://us.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Total trace metal impurities ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR