Sodium bromide 99+% for biochemistry

AC44680-5000 AC44680-1000
103516-822EA 108 USD
103516-822 103516-820
Sodium bromide 99+% for biochemistry
Sodium bromide
Formula: NaBr
MW: 102.89 g/mol
Boiling Pt: 1390 °C (760 mmHg)
Melting Pt: 755 °C
Storage Temperature: Ambient
MDL Number: MFCD00003475
CAS Number: 7647-15-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration Argentometric ≥99.0 %
pH 5.0 to 8.8
Chloride (Cl) ≤0.2 %
Bromate (BrO3) ≤0.001 %
Sulfate (SO4) ≤0.002 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.1 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %
Nitrogen compounds (as N) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR