Sodium borohydride ≥99%, granules, VenPure™ SF, pure

Danger

Synonyms: Sodium tetrahydridoborateSodium tetrahydroborate

AC44850-1000
103528-984EA 73.3 USD
103528-984
Sodium borohydride ≥99%, granules, VenPure™ SF, pure
Sodium borohydride
Formula: NaBH₄
MW: 37.83 g/mol
Melting Pt: 360 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003518
CAS Number: 16940-66-2
UN: 1426
ADR: 4.3,I

Order Now

Specification Test Results

Appearance White Granules
Assay ≥98.5 % (Hydrogen evolution)
Iron (Fe) ≤5 ppm
Silicon dioxide (SiO2) 21 to 500 ppm
Fines ≤8.5 % (< 0.5 cm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR