Silicone oil high temperature
Synonyms:
Polyphenyl-methylsiloxane
AC17466-5000
200001-796EA
245
USD
200001-796
Silicone oil high temperature
Silicone oil
Density:
1.07 g/cm³ Flash Pt: 250 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132673 CAS Number: 63148-58-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...