Silicon dioxide 99.998% (trace metals basis)
|
Warning
|
Synonyms:
Silicon(IV) oxide
, Silicium (IV) oxide
, Silicium dioxide
AC43715-1000
AC43715-0200
AC43715-0050
103515-878EA
48.78
USD
103515-878
103515-882
103515-880
Silicon dioxide 99.998% (trace metals basis)
Silicium dioxide
Formula:
SiO₂ MW: 60.08 g/mol Melting Pt: 1610 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 14808-60-7 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Assay | 44.5 to 47.9 % (Si) |
Total trace metal impurities | ≤20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...