Salicylideneaniline 97%

Warning

Synonyms: 2-((Phenylimino)methyl)phenol

AC33898-0250
103466-464EA 118.8 USD
103466-464
Salicylideneaniline 97%
Salicylideneaniline
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Melting Pt: 49.4…52.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00020066
CAS Number: 779-84-0

Order Now

Specification Test Results

Appearance White to yellow to tan Crystalline powder
Infrared spectrum Conforms
Melting point 47 to 53 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR