Salicylohydroxamic acid 99%
|
Warning
|
Synonyms:
N,2-Dihydroxybenzamide
, Salicylhydroxamic acid
AC13262-0250
AC13262-0050
103466-458EA
96.6
USD
103466-458
103466-456
Salicylohydroxamic acid 99%
Salicylohydroxamic acid
Formula:
C₇H₇NO₃ MW: 153.14 g/mol Melting Pt: 177…185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002110 EINECS: 201-934-3 |
Specification Test Results
Appearance | Pink-beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.5 % |
Elemental Analysis | 8.67 to 9.11 % N |
Specific optical rotation | +6.5° to +9.5° (20 °C, 589 nm) c=5 in 1N HCl |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...