Rhodium(II) trifluoroacetate dimer

Warning

Synonyms: Trifluoroacetic acid rhodium(II) salt dimer

AC38970-0010 AC38970-2500
103518-798EA 98 USD
103518-798 103518-796
Rhodium(II) trifluoroacetate dimer
Rhodium(II) trifluoroacetate dimer
Formula: C₈F₁₂O₈Rh₂
MW: 657,87 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209611
CAS Number: 31126-95-1

Order Now

Specification Test Results

Appearance Green Crystalline powder
Infrared spectrum Conforms
Rhodium (Rh) 27.5 to 35.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR