5-Quinoxalinecarboxylic acid 95%

Warning

Synonyms: Quinoxaline-5-carboxylic acid

AC42958-0010 AC42958-0050
103466-440EA 529.3 USD
103466-440 103466-438
5-Quinoxalinecarboxylic acid 95%
5-Quinoxalinecarboxylic acid
Formula: C₉H₆N₂O₂
MW: 174.1561 g/mol
Melting Pt: 167.6…171.7 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD02854494
CAS Number: 6924-66-9

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Melting point 166 to 173 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR