Quinine sulfate dihydrate, meets analytical specification of USP

Warning

Synonyms: bis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulphuric acid dihydrate(-)-Quinine sulfate dihydrate(-)-Quinine sulphate dihydratebis((R)-[(2S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate

AC41878-0050 AC41878-1000 AC41878-0250
103514-882EA 363.68 USD
103514-882 103514-884 103514-880
Quinine sulfate dihydrate, meets analytical specification of USP
(-)-Quinine sulphate dihydrate
Formula: C₄₀H₅₀N₄O₈S·2H₂O
MW: 782.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150790
CAS Number: 6119-70-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR