Quartz, fine wool 4 µm

Warning

Synonyms: Sand, white quartzSandSilicium (IV) oxideSilicon dioxideSilicium dioxide

AC45104-0500 AC45104-0100
103518-170EA 147 USD
103518-170 103518-168
Quartz, fine wool 4 µm
Quartz
Formula: SiO₂
MW: 60.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00217788
CAS Number: 14808-60-7
https://us.vwr.com/stibo/web/std.lang.all/05/83/16640583.jpg

Order Now

Specification Test Results

Appearance White Fibers

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR