Pyridoxamine dihydrochloride 98%

Warning

Synonyms: 4-(Aminomethyl)-5-hydroxy-6-methyl-3-pyridinemethanol dihydrochloride

AC43625-0050 AC43625-0010
103515-780EA 69.99 USD
103515-780 103515-782
Pyridoxamine dihydrochloride 98%
Pyridoxamine dihydrochloride
Formula: C₈H₁₂N₂O₂·2HCl
MW: 241.12 g/mol
Melting Pt: 216…218 °C
Storage Temperature: Freezer
MDL Number: MFCD00012808
CAS Number: 524-36-7

Order Now

Specification Test Results

Appearance White to yellow to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 to ≤102.5 % (on dry basis)
Water ≤10 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR