Pumice stone, granular (∼ 1-5 mm)

AC39004-2500 AC39004-0010
103518-800EA 216.46 USD
103518-800 103518-802
Pumice stone, granular (∼ 1-5 mm)
Pumice stone
Storage Temperature: Ambient
MDL Number: MFCD00148147
CAS Number: 1332-09-8

Order Now

Specification Test Results

Appearance Grey-brown Granules
Particle size ≥90 % (0.8 - 3 mm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR