p-Toluidine 99%, crystalline molten mass

Danger

Synonyms: 4-Toluidine4-Aminotoluene4-Methylaniline

AC37635-0250 AC37635-0050 AC37635-2500 AC37635-0010
103528-018EA 203 USD
103528-018 103528-016 103528-022 103528-020
p-Toluidine 99%, crystalline molten mass
p-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 200 °C (760 mmHg)
Melting Pt: 41…46 °C
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00007906
CAS Number: 106-49-0
EINECS: 203-403-1
UN: 3451
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Colorless to red Liquid after melting or solid (May discolor to brown on storage)
Infrared spectrum Conforms
Melting point 41 to 46 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR