Propionic acid ACS

Danger

Synonyms: Propanoic acid

AC44723-1000 AC44723-5000
103524-588EA 81.2 USD
103524-588 103524-586
Propionic acid ACS
Propionic acid
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 141 °C (760 mmHg)
Melting Pt: –22 °C
Density: 0.99 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00002756
CAS Number: 79-09-4
UN: 3463
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.15 %
Color scale ≤20 APHA
Residue after evaporation ≤0.01 %
Titration with NaOH ≥99.5 %
Heavy metals (as Pb) ≤0.001 %
Carbonyl compounds ≤0.002 %
Oxidisable substance (HCOOH) ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR