Potassium wolframate 94% -100 mesh
|
Warning
|
Synonyms:
Potassium tungsten oxide
, Potassium tungstate
AC44687-1000
AC44687-0250
103516-680EA
76.4
USD
103516-680
103516-682
Potassium wolframate 94% -100 mesh
Potassium wolframate
Formula:
K₂O₄W MW: 326.03 g/mol Melting Pt: 921 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049668 CAS Number: 7790-60-5 |
Specification Test Results
Appearance | White Powder |
Assay | 53 to 56 % (W) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...