Potassium molybdate(VI) 95%
|
Warning
|
Synonyms:
di-Potassium molybdate
, Dipotassium tetraoxomolybdate
, Potassium molybdate
, Molybdic acid potassium salt
, Potassium molybdenum oxide
, Molybdic acid, potassium salt
AC39323-1000
AC39323-5000
103517-888EA
217
USD
103517-888
103517-886
Potassium molybdate(VI) 95%
Potassium molybdate(VI)
Formula:
K₂MoO₄ MW: 238.13 g/mol Melting Pt: 919 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011370 CAS Number: 13446-49-6 |
Specification Test Results
Appearance | White Powder and/or chunks |
Assay | ≥94 % |
Chloride (Cl) | ≤0.05 % |
Phosphate (PO4) | ≤0.03 % |
Sulfate (SO4) | ≤0.05 % |
Iron (Fe) | ≤0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...