Potassium hydroxide 99.98% (trace metals basis), pellets

Danger

Synonyms: Potassium hydrateCaustic potash

AC43713-5000 AC43713-1000
103528-882EA 338 USD
103528-882 103528-880
Potassium hydroxide 99.98% (trace metals basis), pellets
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320 °C (760 mmHg)
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8,II

Order Now

Specification Test Results

Appearance White Pellets
Total trace metal impurities ≤200 ppm
Titration with HCl ≥59.5 % (K)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR