Potassium hydroxide ∼50% in water

Danger

Synonyms: Potassium hydrateCaustic potash

AC39606-0025
103528-458EA 102.82 USD
103528-458
Potassium hydroxide ∼50% in water
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Density: 1.51 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless Viscous solution
Titration with HCl 49 to 51 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR