Potassium hydrogen carbonate ACS

Synonyms: Potassium bicarbonate

AC44630-5000
103518-946EA 59.3 USD
103518-946
Potassium hydrogen carbonate ACS
Potassium hydrogen carbonate
Formula: KHCO₃
MW: 100.12 g/mol
Melting Pt: 292 °C
Storage Temperature: Ambient
MDL Number: MFCD00011402
CAS Number: 298-14-6

Order Now

Specification Test Results

Appearance Colorless to white Crystals or crystalline powder
Titration with HCl 99.7 to 100.5 % (On dried basis)
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤5 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.03 %
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR