Potassium ethyltrifluoroborate 97%
|
Warning
|
Synonyms:
Potassium ethyltrifluoroboranuide
AC44488-0050
AC44488-0010
103518-934EA
96.5
USD
103518-934
103518-936
Potassium ethyltrifluoroborate 97%
Potassium ethyltrifluoroborate
Formula:
C₂H₅BF₃K MW: 135.9656 g/mol Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112713 CAS Number: 44248-07-9 |
Specification Test Results
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | ≥260 °C |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...