Potassium carbonate, anhydrous 98%, Extra Pure

Warning

Synonyms: Carbonic acid potassium saltCarbonic acid, dipotassium salt

AC44762-0050 AC44762-0010
103516-920EA 110 USD
103516-920 103516-918
Potassium carbonate, anhydrous 98%, Extra Pure
Potassium carbonate
Formula: K₂CO₃
MW: 138.21 g/mol
Melting Pt: 891 °C
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
https://us.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤2.0 %
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR