Potassium bromide for spectroscopy

Warning

AC45732-2500 AC45732-0025
103518-472EA 766 USD
103518-472 103518-474
Potassium bromide for spectroscopy
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (760 mmHg)
Melting Pt: 734 °C
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
Suitability (for IR spectroscopy) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR