Potassium hydrogen sulfate 99%, Extra Pure
|
Danger
|
Synonyms:
Potassium bisulphate
, Potassium bisulfate
AC44761-0025
AC44761-5000
103524-624EA
42.6
USD
103524-624
103524-620
Potassium hydrogen sulfate 99%, Extra Pure
Potassium hydrogen sulphate
Formula:
KHSO₄ MW: 136.17 g/mol Melting Pt: 214 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011404 CAS Number: 7646-93-7 UN: 2509 ADR: 8,II |
Specification Test Results
Appearance | White Crystalline powder |
Solubility | (10 % in water) Clear colorless |
Titration with NaOH | ≥98.5 % |
Chloride (Cl) | ≤0.002 % |
Ammonium (NH4) | ≤0.003 % |
Iron (Fe) | ≤0.003 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...