Polyvinyl alcohol, M.W. ∼ 146,000-186,000 92-94%, hydrolysed

Synonyms: Mowiol®PVOHPVA

AC18316-1000 AC18316-0025
103463-778EA 28.65 USD
103463-778 103463-776
Polyvinyl alcohol, M.W. ∼ 146,000-186,000 92-94%, hydrolysed
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
Melting Pt: 230…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR