Poly(DL-lactide-co-glycolide), M.W. ∼ 19,000 ( Lactide:Glycolide) (50:50)%

Synonyms: PLGA

AC43620-0050 AC43620-0010
103515-778EA 422.15 USD
103515-778 103515-776
Poly(DL-lactide-co-glycolide), M.W. ∼ 19,000 ( Lactide:Glycolide) (50:50)%
Poly(DL-lactide-co-glycolide)
Formula: [C₃H₄O₂]x[C₂H₂O₂]y
Storage Temperature: Ambient
MDL Number: MFCD00131930
CAS Number: 26780-50-7

Order Now

Specification Test Results

Appearance White to tan Crystalline powder or granules or chunks
Viscosity 0.15 to 0.25 dl/g
Polymer 45 to 55 % (Lactide)
Polymer 45 to 55 % (Glycolide)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR