Poly(acrylamide), carboxyl modified, M.W. ∼ 100,000, high carboxyl content

Synonyms: 2-Propenamide, polymer with 2-propenoic acid and sodium 2-propenoate

AC44853-2500 AC44853-0050
103466-328EA 157.23 USD
103466-328 103466-326
Poly(acrylamide), carboxyl modified, M.W. ∼ 100,000, high carboxyl content
Poly(acrylamide), carboxyl modified
Storage Temperature: Ambient
MDL Number: MFCD00084393
CAS Number: 62649-23-4

Order Now

Specification Test Results

Appearance Off-white Powder
Infrared spectrum Conforms
Loss on drying ≤7.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR