PIPES disodium salt 99%

Synonyms: Disodium piperazine-1,4-diethanesulfonatePiperazine-N,N'-bis-2-ethanesulphonic acid disodium saltPiperazine-1,4-bis(2-ethanesulfonic acid) disodium saltDisodium piperazine-1,4-diethanesulphonate2,2'-(Piperazine-1,4-diyl)bis(ethanesulfonic) acid disodium salt1,4-Piperazinediethanesulphonic acid disodium saltPIPES-Na21,4-Piperazinediethanesulfonic acid disodium saltPiperazine-1,4-bis(2-ethanesulphonic acid) disodium salt2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid disodium saltPiperazine-N,N'-bis-2-ethanesulfonic acid disodium salt

AC21509-2500 AC21509-1000
103466-960EA 346 USD
103466-960 103466-958
PIPES disodium salt 99%
PIPES disodium salt
Formula: C₈H₁₆N₂Na₂O₆S₂
MW: 346.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064349
CAS Number: 76836-02-7

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Water ≤5 %
Solubility (20 % in water) Clear colorless
UV (0.1 M in water)
UV at 260 nm A: ≤0.050
UV at 280 nm A: ≤0.050
pH 9.2 to 10.0 (1 % in water at 25 °C)
Titration with HCl 98.5 to 101.5 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR